Would you like to log in again?
PTFE-Lined Closed Caps, Chemglass
Supplier: Chemglass
CG-895-07 CG-895-04 CG-895-02 CG-895-03
10170-784PK 179.51 USD InStock
10170-784 10170-782 10170-362 10170-780
PTFE-Lined Closed Caps, Chemglass
Closures Bottle Closures
Caps features foam backed PTFE that resists attack from virtually all chemicals at room temperature, while providing a leak-tight seal.
  • Temperature tolerant
  • Not autoclavable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty