Would you like to log in again?
smartSpatulas™ Disposable Spatulas, Chemglass
Supplier: Chemglass
CG-1985-P-07 CG-1985-P-05 CG-1985-P-01 CG-1985-P-03
10170-386PK 57.59 USD InStock
10170-386 10170-384 10170-382 10170-380
smartSpatulas™ Disposable Spatulas, Chemglass
Spatulas
Look for the V at the neck.™ Versatile polypropylene tools provide endless utility in the lab, including transferring, handling, and manipulating a variety of chemicals and biological materials.
  • Polypropylene is autoclavable, resistant to dilute acids and bases, and won’t shatter after exposure to liquid nitrogen frozen samples
  • Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer
  • Double-ended design to scoop, scrape, spoon, stir, collect, transfer, handle or manipulate a variety of biological or chemical materials
  • SGS-Tested for US and EU food contact standards
  • Available EtO sterilized
Multiple sizes for diverse applications. Antistatic micro for manipulating extremely small quantities and fits into 0.2 ml tubes. Utilitarian standard fits into 0.5 ml and 15 ml tubes. Macro allows easier access to large containers, has a clump busting pick, and fits into 15 ml and 50 ml tubes. Short is useful in smaller spaces and fits into 0.5 ml and 15 ml tubes. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc. US Patent 8,709,362. Made in China.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty