Would you like to log in again?
Septum Stoppers, Sleeve Type, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-3022-09 CG-3022-95 CG-3022-96 CG-3022-89 CG-3022-88 CG-3022-99 CG-3022-97 CG-3022-91 CG-3022-93 CG-3022-98
89029-091PK 53.73 USD InStock
89029-091 89029-092 33501-388 33501-630 10170-488 10170-486 89029-089 10170-492 80062-432 10170-490
Septum Stoppers, Sleeve Type, Chemglass
Stoppers Rubber Stoppers
Sleeve type stoppers with hollow plugs and sleeve extensions.
  • Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty