Would you like to log in again?
KIT AMBER WITH SPOT 2ML 12X32MM 9MM
Supplier: Chemglass
Kit, Amber with Spot 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040in Red PTFE/Silicone Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT AMBER WITH SPOT 2ML 12X32MM 9MM 10170-590 CV-5150-1232 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty