Would you like to log in again?
Wisp™ Style, Amber and Clear Vials, Chemglass
Supplier: Chemglass
WISP™
CV-1259-1545 CV-1258-1545 CV-1257-1545 CV-1256-1545
75820-640PK 53.92 USD InStock
75820-640 10170-630 75820-638 89493-008
Wisp™ Style, Amber and Clear Vials, Chemglass
Vials
This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers.
  • Clear or amber borosilicate
  • Available with graduated marking spots
These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty