Would you like to log in again?
ChemRxnHub™ Systems, Jacketed, 500 ml, Jacketed Lab Reactor, Chemglass
Supplier: Chemglass
CG-1949-X-500 CG-1949-X-500M CG-1949-X-505M CG-1949-X-505
10170-678EA 17167.97 USD InStock
10170-678 10171-106 30622-582 30622-584
ChemRxnHub™ Systems, Jacketed, 500 ml, Jacketed Lab Reactor, Chemglass
Laboratory Reactor Systems
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty