Would you like to log in again?
DURAN® Media Storage Bottles with Hose Barb, Clear Glass, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLS-1423-500 CLS-1423-5L CLS-1423-100 CLS-1423-2L CLS-1423-10L CLS-1423-1L CLS-1423-250
10170-338EA 642.34 USD InStock
10170-338 10170-778 10170-776 10170-336 10170-334 10170-332 10170-340
DURAN® Media Storage Bottles with Hose Barb, Clear Glass, Chemglass
Bottles Cell Culture Bottles
Use these clear Duran® media bottles for easy gravity transfer of solutions.
  • Drip-free pouring ring
  • One-piece, liner-less vented cap is autoclavable and reusable
The hose barb outlet at the bottom allows for easy gravity transfer of solutions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty