Would you like to log in again?
Screw Thread Vials, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CV-1117-1232 CV-1119-1232 CV-2595-1232 CV-2600-1232 CV-2605-1232 CV-1120-1232 CV-2590-1232 CV-1116-1232
75820-396PK 39.38 USD InStock
75820-396 10170-624 75820-394 10170-622 10171-026 75820-398 10171-022 75820-392
Screw Thread Vials, Chemglass
Vials
Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa.
  • Available with graduated marking spots
  • Vials are available in silanized form
This vial is designed to work in a variety of auto samplers requiring narrow neck vials.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty