Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Epredia™ Blue Nylon Biopsy Bags, Epredia
Supplier: Epredia
Epredia™
6774011 6774015 6774015 6774014 6774010 6774013 6774009 6774012
10170-218CS 814.11 USD InStock
10170-218 10171-080 10171-078 CA89212-290 89212-290 10170-220 10171-076 10171-082
Epredia™ Blue Nylon Biopsy Bags, Epredia
Bags Sample Bags
Ensure unrestricted fluid movement around tissue with Epredia™ Blue Nylon biopsy bags.
  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty