Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Supplier: Thermo Scientific
Nunclon™
150288 168381 153066 169558 171099 157150 174888 150318 150350 172931 150288 150326 153066 168381 171099 174888 157150 174926 169558 172931 166508 174926 150350 166508 150326 150318
25387-052CS 691.43 USD InStock
CA25382-337 CA25382-336 25387-052 CA25382-335 25382-327 CA25382-334 CA25382-332 CA25382-331 CA25382-330 CA62407-376 62407-124 CA10769-140 CA10171-744 10753-500 CA25382-327 25382-337 25382-335 25382-336 25382-334 25382-331 25382-332 25382-330 10171-744 CA25387-052 CA62407-124 62407-376
Nunc™ Cell Culture/Petri Dishes, Thermo Scientific
Culture Dishes Cell Culture Dishes
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs with Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes
These cell culture/petri dishes are made of polystyrene.
These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.
Ordering information: Quality certificates are included in each case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty