Would you like to log in again?
Follicle Dermal Papilla Cells (HFDPC), PromoCell
Supplier: PromoCell
C-14005 C-12072 C-12071
10171-904EA 1154.34 USD InStock
10171-904 10172-172 10171-902
Follicle Dermal Papilla Cells (HFDPC), PromoCell
Cells
Primary Human Follicle Dermal Papilla Cells (HFDPC) are isolated from human dermis originating from the lateral scalp. Information on donor hair and skin color is available for each lot. HFDPC stain positive for alkaline phosphatase.
  • Alkaline phosphatase positive
  • Cryopreserved at second passage
  • Available from adult lateral scalp
  • Available from donors with different hair colors
Dermal papillae are embedded in a laminin and collagen IV rich extracellular matrix at the base of the hair follicles
This product can be used in the following applications. Hair growth research, Drug testing, Toxicological testing.
They are essential for the induction and maintenance of hair growth. Since they have androgen receptors, they can be used for in vitro screening of androgen blocking reagents.
Ordering information: Human Follicle Dermal Papilla Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty