Would you like to log in again?
Human umbilical artery endothelial cells (HUAEC), PromoCell
Supplier: PromoCell
C-12202 C-14013 C-12252
10171-908EA 769.54 USD InStock
10171-908 10175-202 10171-958
Human umbilical artery endothelial cells (HUAEC), PromoCell
Cells
Primary Human Umbilical Artery Endothelial Cells (HUAEC) are isolated from the arteries of the umbilical cord and are often used for in vitro studies on atherosclerosis but also for endothelial cell-dependent lymphocyte activation and chemoattractant activity experiments
  • Von Willebrand factor (vWF) positive
  • CD31 positive, Dil-Ac-LDL uptake positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
(Human Umbilical Vein Endothelial Cells (HUVEC) from the same donor are available on request).
Ordering information: Human Umbilical Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty