Would you like to log in again?
Human Dermal Lymphatic Endothelial Cells (HDLEC), PromoCell
Supplier: PromoCell
C-12216 C-14020 C-12217 C-12218 C-14021 C-12219
10171-932EA 1142.61 USD InStock
10171-932 10172-182 10171-938 10172-180 10171-934 10171-936
Human Dermal Lymphatic Endothelial Cells (HDLEC), PromoCell
Cells
Primary Human Dermal Lymphatic Endothelial Cells (HDLEC) are a subpopulation of the Human Dermal Endothelial Cells. They are isolated from the dermis of juvenile foreskin and adult skin (different locations) from a single donor.
(Human Dermal Blood Endothelial Cells (HDBEC) from the same donor are available on request.) The cells are routinely analyzed by flow cytometric analysis and immunofluorescent staining: > 95% of the cells are CD31 positive  and podoplanin positive. In addition, the cells stain positive for Prox-1.
Lymphatic vessels transport excess fluids from tissues to the circulatory system and are a major component of the immune system
This product can be used in the following applications. Angiogenesis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
The transported fluid is nearly cell free. The vessels are highly permeable, because they lack a continuous basal membrane. Lymphatic endothelial cells are involved in pathological alterations of the lymphatic system. During tumor lymphangiogenesis, the lymphatic endothelial cells build new vessels that infiltrate tumors, attract tumor cells, and induce tumor cell metastasis.
Ordering information: Human Dermal Blood Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty