Would you like to log in again?
Human saphenous vein endothelial cells (HSaVEC), PromoCell
Supplier: PromoCell
C-12231 C-14025 C-12232
10171-950EA 1187.05 USD InStock
10171-950 10171-948 10172-190
Human saphenous vein endothelial cells (HSaVEC), PromoCell
Cells
Primary Human Saphenous Vein Endothelial Cells (HSaVEC) are isolated from human saphe-nous veins from single donors
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
The saphenous vein is often used for autotransplantation in coronary artery bypass operations, when arterial grafts are not available or many grafts are required.
Ordering information: Human Saphenous Vein Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty