Would you like to log in again?
Human Tracheal Smooth Muscle Cells (HTSMC), PromoCell
Supplier: PromoCell
C-14056 C-12565 C-12566
10172-106EA 987.07 USD InStock
10172-106 10172-104 10172-230
Human Tracheal Smooth Muscle Cells (HTSMC), PromoCell
Cells
Primary Human Tracheal Smooth Muscle Cells (HTSMC) are isolated from the trachea from single donors and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
They connect the endings of the tracheal cartilage by a contractile bridge called the paries membranaceus and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the trachea walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HTSMC are suitable for in vitro studies of asthma and other pulmonary diseases.
Ordering information: Human Tracheal Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty