Would you like to log in again?
Human Uterine Smooth Muscle Cells (HUtSMC), PromoCell
Supplier: PromoCell
C-14059 C-12576 C-12575
10172-116EA 1178.09 USD InStock
10172-116 10172-236 10175-144
Human Uterine Smooth Muscle Cells (HUtSMC), PromoCell
Cells
Primary Human Uterine Smooth Muscle Cells (HUtSMC) are isolated from the myometrium (the middle layer of the uterine wall) and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
During pregnancy, hormones and growth factors cause an extensive hypertrophy of uterine smooth muscle cells. This and other morphological and biochemical effects on the uterus can be studied in vitro using HUtSMC.
Ordering information: Human Uterine Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty