Would you like to log in again?
Human Pericytes from Placenta (hPC-PL), PromoCell
Supplier: PromoCell
C-14095 C-12980 C-12981
10172-160EA 980.58 USD InStock
10172-160 10175-212 10172-162
Human Pericytes from Placenta (hPC-PL), PromoCell
Cells
Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls. They are important for angiogenesis, the structural integrity of the microvasculature, and blood flow regulation. However, they can also develop into malignant tumors called hemangiopericytomas.
To date, pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells, and phagocytes (macrophages).
This product can be used in the following applications. Angiogenesis research, Regenerative medicine research, Oncology
Ordering information: Human Pericytes from human placenta can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty