Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride 0.5 M in water

Supplier: Promocell, Inc.
Synonyms: Chloro calcium

C-34006
10172-424EA 66.78 USD InStock
10172-424
Calcium chloride 0.5 M in water
Calcium chloride solutions
Calcium chloride solution allows the adjustment of the optimal amount of calcium required by the cells. Calcium chloride (CaCl₂) solution is prepared as a solution from highly pure tissue culture tested calcium chloride and cell culture-grade water.

  • Clear, colorless and sterile solution

Some types of primary human cells, especially normal human epidermal keratinocytes (NHEK), respond sensitively to the calcium concentration in the culture medium.

Caution: Store at -20°C in the dark immediately after arrival. The products are for in vitro research use only and not for diagnostic or therapeutic procedures.


Formula: CaCl₂·2H₂O
Storage Temperature: Freezer
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty