Would you like to log in again?
HEPES Buffered Saline, Promocell
Supplier: PromoCell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C-40000 C-40010 C-40020
10172-534EA 17.23 USD InStock
10172-534 10172-538 10172-536
HEPES Buffered Saline, Promocell
Buffers Cell Culture Buffers
These solutions are prepared from raw materials of the highest quality using tissue culture grade water.
  • Store at –20 °C
  • Ready-to-use
Used in tissue culture, cell isolation procedures, and molecular biology.
Caution: For in vitro research use only and not for diagnostic or therapeutic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty