Would you like to log in again?
Serum Testosterone ELISA
Supplier: ALPCO
11-TESHU-E01
101720-134EA 313.85 USD InStock
101720-134
Serum Testosterone ELISA
Assays ELISAs
The Serum Testosterone ELISA is for the direct quantitative determination of testosterone by enzyme immunoassay in human serum.
Testosterone is the most important male sex hormone and is responsible for genital development, beard growth, muscle development, and general male characteristics. The measurement of serum or plasma levels is an index of Leydig cell function and high or low values correlate well with hypo- or hypergonadism. In females, the adrenals and ovaries produce small amounts of testosterone. High levels of testosterone in females indicate excessive androgen production and are found in progressive hirsutism and virilization, Cushing’s syndrome, and a deficiency in one or more of the specific enzymes required for normal steroid biosynthesis.
Certifications: FDA Registered.
Caution: For in vitro diagnostic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty