Would you like to log in again?
DHEAS ELISA
Supplier: ALPCO
11-DHEHU-E01
101720-136EA 312.31 USD InStock
101720-136
DHEAS ELISA
Assays ELISAs
The Dehydroepiandrosterone Sulfate ELISA is intended for the direct quantitative determination of DHEA-S by enzyme immunoassay in human serum.
Dehydroepiandrosterone sulfate (DHEAS) is produced by the adrenals and gonads. As a result, the determination of the level of DHEAS in serum is important in the evaluation of the functional state of these glands. DHEAS is a precursor of testosterone and estrone. Besides the adrenals in females, the ovaries have been shown to be an important source of DHEAS. It has been reported that there is a fluctuation day by day of DHEAS in women during the ovulatory cycle. The principle production of testosterone in females is from conversion of other related androgens, especially DHEAS. An abnormal testosterone level in women should be accompanied by the estimation of serum DHEAS. The use of serum testosterone determination in conjunction with the ELISA of DHEAS can be used to determine if the source of excess androgen production is ovarian or adrenal.
Certifications: FDA Registered.
Caution: For In Vitro diagnostic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty