Would you like to log in again?
Progesterone ELISA
Supplier: ALPCO
11-PROHU-E01
101720-142EA 332.31 USD InStock
101720-142
Progesterone ELISA
Assays ELISAs
The Progesterone ELISA is for the direct quantitative determination of Progesterone by enzyme immunoassay in human Serum.
  • FDA registered; for in vitro diagnostic use
Progesterone is a C-21 female sex steroid hormone with a variety of physiological effects. In the follicular phase of the menstrual cycle, progesterone is produced in low levels. It increases to the LH peak and then sharply rises 3 to 4 days later to higher levels, remaining elevated through the 10th to 12th days after the LH peak. Then there is a sharp decline to the low levels of the follicular phase. It is responsible for the induction of the cyclic changes in the endometrium of the uterus allowing implantation and successful growth of the fertilized ovum and maintenance of pregnancy.
Progesterone measurements are used in documenting ovulation and in the management of difficulties during the first trimester of pregnancy. Levels of progesterone may be useful in the evaluation of sterility due to luteal phase defects, prediction of impending abortion, and the diagnosis of ectopic pregnancy. Normal values of progesterone may be affected by drugs such as, oral contraceptives, super ovulatory drugs, estrogen replacement therapy medication, and GnRH analogues. The removal of ovarian function following surgical oophorectomy or chemotherapy may influence serum progesterone values.
Certifications: FDA registered. For in vitro diagnostic use in the United States.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty