Would you like to log in again?
Cortisol ELISA (Saliva)
Supplier: ALPCO
11CORHUE01SLV
101720-168EA 332.31 USD InStock
101720-168
Cortisol ELISA (Saliva)
Assays ELISAs
Cortisol ELISA (Saliva) is an enzyme immunoassay for the quantitative determination of cortisol in human saliva. For research use only. Not for use in diagnostic procedures.
  • Includes ready to use standards and controls
  • No cross reaction with DHEAS and tetrahydrocortisone
  • Incubation time of 60 to 65 minutes
Cortisol is the most abundant circulating steroid and the major glucocorticoid secreted by the adrenal cortex. Studies have shown cortisol is physiologically effective in blood pressure maintenance and anti-inflammatory activity. It is also involved in calcium absorption, gluconeogenesis and the secreation of gastric acid and pepsin.
Most circulating cortisol is bound to cortisol binding protein, transcortin, or albumin. In the absence of cortisol binding proetins in saliva, salivary cortisol is concidered to be free and may show a diurnal rhythm with the highest levels in the morning an the loweest levels at night.
Research ares interests include adrenal function, addisons and cushings deseases, adrenal hyperplasia, carcinoma, and stress studies.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty