Would you like to log in again?
5α-Androstane-3α, 17ß-Diol Glucuronide ELISA
Supplier: ALPCO
11-ANDHU-E01
101720-200EA 395.38 USD InStock
101720-200
5α-Androstane-3α, 17ß-Diol Glucuronide ELISA
Assays ELISAs
For the direct quantitative determination of 3α-Diol G by enzyme immunoassay in human serum.
5α-Androstane-3α, 17β-diol glucuronide is a C19 steroid and is either abbreviated as 3α-Diol G, 5α-Diol G or simply, α-Diol G. It is produced mainly as a metabolite of testosterone and dihydrotestosterone (DHT). It is largely produced in target peripheral tissues such as the skin, especially around hair follicles.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty