Would you like to log in again?
Leptin ELISA
Supplier: ALPCO
11-LEPHU-E01
101720-304EA 543.08 USD InStock
101720-304
Leptin ELISA
Assays ELISAs
The Leptin ELISA is for the quantitative determination of leptin in human serum.
  • Analytical sensitivity 0.42 ng/ml
  • Dynamic range 1 to 100 ng/ml
The Leptin ELISA follows a typical two-step sandwich type assay format. The Leptin ELISA makes use of two highly specific monoclonal antibodies: A monoclonal antibody specific for leptin is immobilized onto the microWells plate and another monoclonal antibody specific for a different epitope of leptin is conjugated to biotin. The Leptin ELISA includes all components (96 well Ab coated plate, conjugate Ab, substrate, detector, standards, quality controls and dilution and wash buffer) necessary to complete the Leptin ELISA.
Caution: Research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty