Would you like to log in again?
Rat C-Peptide ELISA Kit, ALPCO
Supplier: ALPCO
80-CPTRT-E01
101720-806EA 678.46 USD InStock
101720-806
Rat C-Peptide ELISA Kit, ALPCO
Assays ELISAs
The Rat C-peptide ELISA quantifies the C-peptide protein products of rat I and rat II proinsulin genes.
  • 10 µl of sample is sufficient for the assay
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty