Would you like to log in again?
ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit, Alpco
Supplier: ALPCO
30-KIF005
101721-416EA 1003.08 USD InStock
101721-416
ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit, Alpco
Assays Cellular Assays
ID-Vit® Folic acid is a microtiter plate test kit based on a microbiological method which measures the total folic acid content in serum.
The test kit contains the standard and all reagents required to perform the test. It is sufficient for 96 determinations including standard curves.
Delivery information: Kit includes Lactobacillus rhamnosus (precoated microtiter plate), 4 x 5 mL sample treatment solution, 4 x 30 mL water, 4 x folic acid assay medium, 4 x folic acid standard, lyoph., 4 x adhesive cover foil, replacement holder for microtiter strips, 4 x 1.5 mL folic acid medium treatment buffer, 4 x folic acid control 1, lyoph. and 4 x folic acid control 2, lyoph.
Caution: For use in human and veterinary medicine and in research. For in vitro diagnostic use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty