Biacore maintenance kits are convenient kits for proper instrument maintenance.
*Not recommended for use with Biacore 4000, Biacore A100 or Biacore S51.
Ordering information: The use of the CAPture kits in Biacore systems requires an Amine Coupling Kit with Sensor Chip CM5 CM4 CM3 or C. Not for use with Biacore Flexchip unless otherwise noted.