Would you like to log in again?
Buffers for Biacore Systems, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BR100354 BR100358 BR100357 BR100356 BR100350 BR100352 BR100355 BR100351 BR100349
101724-650EA 201.32 USD InStock
101724-650 101724-642 101724-632 101724-644 101724-634 101724-646 101724-636 101724-648 101724-638
Buffers for Biacore Systems, Cytiva
Buffers Assay Buffers
Buffers developed and verified to work in Biacore™ systems.
Immobilisation buffers
A range of buffers for the most common ligand types and immobilisation conditions.
Regeneration buffers
Regeneration is the step where bound analyte is removed from the sensor chip after analysis, without affecting the activity of the immobilised ligand. In many systems, conditions that remove analyte tend to reduce ligand activity, and finding the optimal conditions is an essential part of assay development.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty