Would you like to log in again?
Buffers, running buffers for Biacore Systems, Cytiva
Supplier: Cytiva
BR100671 BR100369 BR100669 BR100828 BR100826 BR100368 BR100188 BR100670 BR100827
101724-784EA 188.51 USD InStock
101724-784 101724-652 101724-752 101724-654 101724-786 101724-754 101724-788 101724-756 101724-604
Buffers, running buffers for Biacore Systems, Cytiva
Buffers Electrophoresis Buffers Running Buffers
Buffers developed and verified to work in Biacore™ systems.
The recommended running buffer for an assay depends on the type of molecules used in the interaction, which assay will be run, and the type of sensor chip used. Supplied in ready to use or concentrated form.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty