Would you like to log in again?
Human Bronchial Epithelial Cells (HBEpC), PromoCell
Supplier: PromoCell
C-14063 C-12641 C-12640
10172-242EA 310.75 USD InStock
10172-242 10175-152 10175-154
Human Bronchial Epithelial Cells (HBEpC), PromoCell
Cells
Primary Human Bronchial Epithelial Cells (HBEpC) are isolated from the surface epithelium of human bronchi and stain positive for cytokeratin
  • Cytokeratin positive
  • Routinely morphology checked
  • Cryopreserved at second passage
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
The respiratory epithelia are responsible for the lubrication of the lungs, the maintenance of humidity, and the cleaning of the respiratory tract. They are an important target for drugs, toxins, and carcinogens. In addition, they are involved in many diseases such as cystic fibrosis. Consequently, HBEpC are useful for investigating the function and pathology of the respiratory system.
Ordering information: Human Bronchial Epithelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty