Would you like to log in again?
Cell Detachment Solutions, DetachKit, PromoCell
Supplier: PromoCell
C-41210 C-41200 C-41220
10175-292EA 70.5 USD InStock
10175-292 10175-300 10175-296
Cell Detachment Solutions, DetachKit, PromoCell
Cell Culture Supplements and Additives
This DetachKit is used for gentle and effective detachment of adherent primary human cells in routine subculturing.
  • Each batch is tested on primary human cells
  • Can be used for primary animal cells and cell lines
  • Comes with a Trypsin/EDTA ratio of 0.04%/0.03%
Each DetachKit consists of three components: HEPES BSS (HEPES buffered balanced salt solution), Trypsin/EDTA solution and TNS (Trypsin Neutralization Solution).
HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty