Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Block Embedding Mold, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

69934-10 69934-20 69934-30
101764-940PK 35.26 USD InStock
101764-940 101764-936 101764-938
Block Embedding Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
Introducing the EM5 - an innovative new design based on the widely used 8 mm Ø size embedding mold.

  • Separate base and tube sections that clip together to form the familiar capsule shape
  • Care should be taken that each component is free of debris before attaching together, to ensure a proper seal.
  • Easy removal of resin blocks achieved by simply pulling the mold apart by hand
  • The mold can produce blocks up to 20 mm in length, however shorter blocks (~14 mm) allow for easier removal. No razor blades or capsule presses are required
  • The detachable cap section allows for easier access and manipulation of a specimen
  • For more accurate specimen alignment a small amount of resin can be cured or partially cured before clipping to the body and completing the resin block. When fully cured they form strong homogeneous blocks
  • Each mold can be clipped to other molds, allowing for the creation of trays of desired size. For volatile resins, another cap section can be clipped on to the top of the mold, creating an air tight seal
  • Each cavity is numbered 15, whilst the end support stands have designated areas for writing indexing numbers or comments

The unique two part design allows for easy removal of resin blocks, as well as incorporating many other advantages over currently available embedding products.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty