Would you like to log in again?
Lighted Forceps, Scissors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77905-03 77905-07 77905-01 77905-05 77905-06 77905-04 77905-02 77905-08
101765-002KT 475.59 USD InStock
101765-002 102098-812 101765-004 101765-006 101764-998 101765-008 101764-996 101765-000
Lighted Forceps, Scissors, Electron Microscopy Sciences
Forceps
See your samples and your embeddings in a whole new light!
Increases the visibility of the specimen
Allows for accurate orientation of tissue and specimen
Minimizers floaters
Reduces embedding and prep time
These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
Delivery information: Each kit includes: one instrument, power pack and battery charger.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty