Would you like to log in again?
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69130-15 69131-03 69130-20 69131-04 69131-01 69130-45 69130-40 69130-10 69131-02 69130-55 69130-25
101765-100EA 1508.27 USD InStock
101765-100 102096-518 102096-528 102096-530 102096-522 101765-094 102096-520 102096-526 101765-096 102096-524 101765-098
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Tissue Embedders
Paraffin tissue microarrays (TMAs) are becoming more common in the laboratories. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process. Arraymold features include:
  • Allow pathologist or researcher to view many different samples on the same single slide
  • Helpful in finding IHC markers for control or quality assurance from hundreds of samples
  • Staining markers will be more consistent when hundreds of samples are on one slide
  • Used less costly antibody
  • Small enough to be stored in a drawer
  • Rubber arraymold can be reused many times without losing its flexibility
Current techniques including organizing, punching, and inserting tissues is time consuming and involving costly equipments.This Arraymold is the simple tool but great answer for this current technique for laboratories to perform TMAs with less time consuming, excellent results. With this Arraymold, you can process up to 150 specimens onto one single slide with very little time consuming, minimal technical training.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty