Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine

Supplier: Matrix Scientific
Synonyms: 2,4,6-Tris(perfluoroethyl)-1,3,5-triazine

002950-1G
101784-964EA 120 USD InStock
101784-964
2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
Mass Spec. Standard

Hazard: Irritant


Formula: C₉F₁₅N₃
MW: 435.10 g/mol
Boiling Pt: 121…122 °C
Density: 1.65
MDL Number: MFCD00042437
CAS Number: 858-46-8
Order Now
Specification Test Results
Boiling point 121-122°C
Density 1.65
ND20 1.3155

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty