Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

Supplier: Matrix Scientific
Synonyms: 2,4,6-Tris(perfluoropropyl)-1,3,5-triazine

002954-1G
101784-968EA 121.54 USD InStock
101784-968
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
Mass Spec. Standard

Hazard: Irritant


Formula: C₁₂F₂₁N₃
MW: 585.12 g/mol
Boiling Pt: 165 °C
Density: 1.71
MDL Number: MFCD00042438
CAS Number: 915-76-4
Order Now
Specification Test Results
Boiling point 165°C
Density 1.71

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty