Would you like to log in again?
Staticmaster® Mini Ionizing Fan, NRD
Supplier: NRD
4068
10183-828EA 256.81 USD InStock
10183-828
Staticmaster® Mini Ionizing Fan, NRD
Ionizers
The NRD 4068 Mini-Fan Ionizer is the perfect solution for objects that have an irregular shape and need to have their charge removed before being used in the balance chamber.
  • Delivers a wide stream of Ions to dissipate static charge over wider area
  • Useful for objects that have an irregular shape
  • Removes charge before use in the balance chamber
One of the biggest challenges in obtaining accurate readings when using a precision balance is the presence of static electricity which can cause weighing errors. While NRD provides solutions for static elimination when weighing fine powders, irregularly shaped objects like plastic weigh boats and spatulas can cause their own set of challenges. Plastic is easily charged in low humidity. Vials and other weighing containers can become charged during delivery, which causes unstable weighing and product "fly-away." Delivering a wide stream of Ions to dissipate the static charge over a wider area, the mini ionizer will complement your other tools in the balance chamber environment.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty