Would you like to log in again?
Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8948
10183-850EA 1919.58 USD InStock
10183-850
Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Micro-Cut Oval Biopsy Punch Forceps are a locking, double-spring plier instrument used for obtaining uterine tissue for biopsy that features razor sharp edges.
The plier handle features a horn for thumb placement as well as a hook and two finger rest to provide maximum hand comfort and greater control of the instrument. The bite of this instrument is the unique Burke-style punch and produces a 3 x 5-mm oval tissue sample. The locking wheel at the base of the shaft has a smooth, beveled side with the rest being cross-serrated which makes the lock easy to engage. This locks the jaws and the tissue sample in place while the instrument is removed from the cervical canal, thus reducing the risk of losing the tissue sample as the instrument is withdrawn.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty