Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Micro-Cut Kevorkain Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8952
10183-852EA 1940.57 USD InStock
10183-852
Micro-Cut Kevorkain Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Micro-Cut Kevorkian Biopsy Punch is commonly used during colposcopy procedures to excise tissue from the endocervix for biopsy
This product has a 3x8mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The Micro-Cut instrument features a knife honed edge on the upper jaw for a cleaner and more precise cut of the tissue. The spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty