Would you like to log in again?
Iowa Puncturing Forceps, OR Grade, Sklar
Supplier: Sklar
91-4310
10184-236EA 393.29 USD InStock
10184-236
Iowa Puncturing Forceps, OR Grade, Sklar
Forceps
Iowa Membrane Puncturing Forceps are an OB/GYN instrument used to puncture the membrane of the amniotic sac and release the amniotic fluid.
They are ratcheted, finger ring forceps with 5 x 6 teeth, that provide a secure hold on the membranes. They have long, narrow shanks and jaws that each have a slight curvature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty