Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Sklar Blue™ Pederson Vaginal Speculum, Straight, OR Grade, Sklar
Supplier: Sklar
91-5034 91-5088 91-5040 91-5092 91-5086 91-5082 91-5032 91-5044 91-5084 91-5036
10184-388EA 493.64 USD InStock
10184-388 10184-444 10184-248 10184-446 10184-450 10184-384 10184-380 10184-250 10184-448 10193-336
Sklar Blue™ Pederson Vaginal Speculum, Straight, OR Grade, Sklar
Specula
Sklar Blue™ Pederson Vaginal Speculum is used for electrocsurgical gynecological procedures, such as LEEP.
  • Sklar Blue instruments are fully autoclavable and may also be cleaned ultrasonically
  • It is a double blade duckbill speculum inserted with the blades closed
  • The blades are then opened by an external lever and screw and more narrow than the blades of the Graves Speculum
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
The practitioner can best determine which type and size speculum is most appropriate for the patient. The Sklar Blue Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty