Would you like to log in again?
Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-9100 90-9106 90-9103 90-9109 90-9102
10184-234EA 1227.93 USD InStock
10184-234 10184-296 10184-286 10184-292 10184-294
Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in laser OB/GYN procedures.
It is available with various blades styles and three weight choices in order to accommodate the needs of the patient and the procedure. The weight holds the speculum in place and provides an open view for the surgeon to perform procedures.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty