Would you like to log in again?
Surgi-OR™ TC Olsen-Hegar Needle Holder, Physician Grade, Sklar
Supplier: Sklar
95-148 95-149
10184-346EA 205.17 USD InStock
10184-346 10184-348
Surgi-OR™ TC Olsen-Hegar Needle Holder, Physician Grade, Sklar
Forceps
The Surgi-OR™ Tungsten Carbide Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built-in scissors.
It is used with intermediate weight sutures, commonly in plastic surgery procedures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty