Would you like to log in again?
Surgi-OR™ Lister Bandage Scissors, Mid-Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgi-OR™
95-232P 95-243 95-232B
10184-352EA 16.34 USD InStock
10184-352 10184-354 10184-350
Surgi-OR™ Lister Bandage Scissors, Mid-Grade, Sklar
Scissors
Surgi-OR™ Lister Bandage Scissors are a versatile instrument with multiple uses
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
They are used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty