Would you like to log in again?
Tischler-Kevorkian Wide Bite Biopsy Punch Forceps, OR Grade, Sklar
Supplier: Sklar
90-9575 90-9570
10184-358EA 1859.83 USD InStock
10184-358 10184-360
Tischler-Kevorkian Wide Bite Biopsy Punch Forceps, OR Grade, Sklar
Forceps
Tischler-Kevorkian Wide Bite Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy.
The jaws of this biopsy punch are designed to open 60 degrees, which is wider than other standard punches, in order to obtain tissue samples from obscure sites. It features a single lower tooth and a single upper tooth to provide a greater grasp on difficult to reach tissue. It has a locking mechansim above the horn on the handle, so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a plier style handle with two finger rests and a hook on the lower handle for user comfort and increased control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty