Would you like to log in again?
Merit™ Mayo Dissecting Scissors, Physician Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
97-312 97-308 97-321 97-325
10193-956EA 14.98 USD InStock
10193-956 10184-408 10193-888 10192-398
Merit™ Mayo Dissecting Scissors, Physician Grade, Sklar
Scissors
Merit™ Mayo Dissecting Scissors are a versatile instrument, that can be used in multiple procedures and multiple settings.
While the curved instrument is used for cutting or dissecting dense or deep tissue, the straight version is used for cutting surface tissue or sutures. Both may be used interchangeably depending on procedure and physician preference.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty