Would you like to log in again?
Merit™ Littauer Scissors, Physician Grade, Sklar
Supplier: Sklar
Merit™
97-358
10184-418EA 19.34 USD InStock
10184-418
Merit™ Littauer Scissors, Physician Grade, Sklar
Scissors
Merit™ Littauer Stitch scissors are used for suture removal.
These littauer scissors have a small hook shaped tip on one blade, that slides under sutures to slightly lift them before cutting for removal. This hook holds the suture, so it does not slip off the blade of the scissors prior to cutting. Littauer Stitch scissors have larger, less delicate tips, and are used for removing larger sutures.
They should not be used for cutting tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty