Would you like to log in again?
Merit™ Weitlaner Retractor, Physician Grade, Sklar
Supplier: Sklar
97-372 97-371 97-376 97-370 97-373
75818-084EA 96.47 USD InStock
75818-084 10193-972 10192-432 10184-426 10184-428
Merit™ Weitlaner Retractor, Physician Grade, Sklar
Retractors
Merit's™ Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site
It has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed. The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries. It is also useful with small, deep incisions and soft tissue dissection at a superficial level. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty