Would you like to log in again?
Surgi-OR™ Ferris Smith Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-449
10184-438EA 90.53 USD InStock
10184-438
Surgi-OR™ Ferris Smith Tissue Forceps, OR Grade, Sklar
Forceps
Surgi-OR™ Ferris Smith Tissue Forceps are heavyweight forceps used with fibrous tissue in colorectal procedures.
These thumb forceps feature a wide section of the handle for greater control handling firm tissue. They also have slightly wider tips that are available with 1 x 2 or 2 x 3 teeth for a more secure grasp of the tissue.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty